HPLC columns, Syncronis™ Silica
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
97005-014001 97002-102130 97005-152130 97005-254630 97005-253030 97003-154630
HYPE97005-014001EA 557 EUR
HYPE97005-014001 HYPE97002-102130 HYPE97005-152130 HYPE97005-254630 HYPE97005-253030 HYPE97003-154630
HPLC columns, Syncronis™ Silica
Kolommen voor chromatografie
Columns for the chromatography of moderately polar organic compounds.
  • Highly pure, high surface area silica
  • Excellent reproducibility for normal phase chromatography
Surface area: 320 m²/g
Carbon load: 0%
pH range: 2 - 8