Zelfklevende pleisters, Salvequick
Leverancier: ORKLA CARE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6943 6470 6454 6036 6444 51030126 51030127
118-0029EA 81.9 EUR
118-0029 118-0030 118-0028 118-0026 118-0027 CEDE51030126 CEDE51030127
Zelfklevende pleisters, Salvequick
Eerste-hulpbenodigdheden Wondverbanden
These plaster refills are available in different versions: Made from plastic or fabric, blue detectable and for sensitive skin.
  • In combination with dispenser: Complete, easy to use system
  • Plasters are always available, ready to use
  • Plasters are allergy-tested
  • Each adhesive bandage is individually wrapped and sterile
Detectable plasters are specially designed for use in the food industry. The plasters are detectable by most metal detectors used within the food industry. The blue colour also makes the plasters easy to see.
Sensitive plasters are white and made of polyurethane. They are water repellent and suitable for sensitive skin.