Single-channel micro pipettes, variable volume, Acura® manual 826 XS
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
826.0010 826.0002 826.0010Y 826.0020 826.0050 826.0100 826.0200 826.1000
613-6864EA 427 EUR
613-6864 613-6863 613-6865 613-6866 613-6867 613-6868 613-6869 613-6870
Single-channel micro pipettes, variable volume, Acura® manual 826 XS
Pipetten Micropipetten
Manual single-channel pipettes with optimised shape/size ratio for superior drivability and improved hand movements when working with micro tubes. Ideal for research laboratories.
  • Short, narrow shaft fitting smaller microtubes
  • Reduced weight (83 to 88 g)
  • Adjustable tip ejector
  • Self-locking click-stops for precise and safe volume setting
  • Autoclavable (fully assembled)
Uniquely tightened lip seal reduces activation forces and provides for ultra-soft pipetting, further reducing hand fatigue while working. Easy user calibration system with integrated key. Locking mechanism protected by calibration seal sticker.
Leveringsinformatie: Individual pipettes are supplied with QC certificate only. The ready to use TwiXS sets include two Acura® manual XS pipettes, one shelve pipette holder, Qualitips® samples, QC certificate and operating instructions.