HPLC columns, Acclaim™ Trinity™ P1
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
071388 071389 071390 071391 075563 075564 075565
555-7208EA 977 EUR
555-7208 555-7209 555-7210 555-7211 555-7481 555-7460 555-7461
HPLC columns, Acclaim™ Trinity™ P1
Kolommen voor chromatografie
The Acclaim™ Trinity™ P1 is a unique, high-efficiency, silica-based column designed for pharmaceutical applications, such as simultaneous separation of pharmaceutical drug substances and their counterions.
  • Adjustable selectivity by changing mobile phase ionic strength, electrolyte type, pH and organic solvent
  • Ideal selectivity for simultaneous separation of basic, neutral, and acidic analytes
  • Retention of ionic and ionisable analytes without using ion-pairing reagents
Particle type: ultra pure, spherical silica
Max. pressure: 4500 psi
Flow rate range: 0,30 to 1,60 ml/min for 3,0 mm ID formats; 0,15 to 0,80 ml/min for 2,1 mm ID formats
Bonded phase: reversed-phase and anion-exchange
pH range: 2,5 to 7,0