Guard columns, EZ-Guard™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP9016 CP9017 CP9019 CP9021 CP9014 CP9013 CP9015 CP9010 CP9011 CP9012 CP9022 CP9023 CP9024 CP9025 CP9026 CP9027 CP9176 CP9177 CP9008
VARICP9016EA 1520 EUR
VARICP9016 VARICP9017 VARICP9019 VARICP9021 VARICP9014 VARICP9013 VARICP9015 VARICP9010 VARICP9011 VARICP9012 VARICP9022 VARICP9023 VARICP9024 VARICP9025 VARICP9026 VARICP9027 VARICP9176 VARICP9177 HEWLCP9008
Guard columns, EZ-Guard™
Kolommen voor chromatografie
EZ-Guard columns combine a FactorFour column with a built-in guard column.
  • Lack of a column connection between the guard and analytical section results in a 100% leak-free column
  • Features a unique uncoated and deactivated outlet section acts as an integrated transfer line
The absence of a stationary phase in the last part of the column significantly reduces background noise. The impact of water, oxygen or other polar or aggressive components that move through the end of the column at high temperature will also be greatly reduced.