HPLC columns, Accucore™ 150-Amide-HILIC
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16726-012105 16726-052130 16726-053030 16726-102130 16726-103030 16726-104630 16726-152130 16726-153030 16726-154630 16726-157569 16726-252130
554-2351EA 705 EUR
554-2351 554-2336 554-2337 554-2339 554-2340 554-2341 554-2342 554-2343 554-2344 554-2346 554-2345
HPLC columns, Accucore™ 150-Amide-HILIC
Kolommen voor chromatografie
These columns are designed for the separation of hydrophilic biomolecules in HILIC mode. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub-2 µm particles.
  • Fast separations with high resolution
  • Low back pressure
  • Recommended for hydrophilic biomolecules such as glycans