IgG purification resin, Pearl™
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-801 786-800
786-801EA 966 EUR
786-801 786-800
IgG purification resin, Pearl™
Eiwitzuivering Protein Purification Resins
The Pearl™ IgG purification resin allows for the one-step purification of immunoglobulin G from serum. The resin binds the high abundant, non-IgG proteins (i.e. albumin) and allows the IgG molecules to pass through in a physiological buffer. The IgG molecules can be stored or used in downstream applications without further clean-up, such as ammonium sulphate precipitation.