SPE columns, CHROMABOND® SB
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
730078 730079 730322 730323 730426 731835 731836 738101.100M 730079.250
MANA730078EA 266 EUR
MANA730078 MANA730079 MANA730322 MANA730323 MANA730426 MANA731835 MANA731836 MANA738101.100M MANA730079.250
SPE columns, CHROMABOND® SB
Kolommen voor chromatografie
CHROMABOND® SB is a quaternary ammonium modified silica anion exchanger (SAX) for SPE.
  • Base material: Silica
  • Strongly basic anion exchanger
Recommended for ion exchange of organic compounds (e.g. organic acids, fatty acids, nucleotides) from aqueous matrix. It is not suited for very strong anions such as sulphonic acids, due to the difficult elution.
Cation exchange capacity: ~0,3 meq/g