Bottles, wide neck, with screw cap
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
80408 80409 80410 80411 80412 80413
VITL80408EA 67.8 EUR
VITL80408 VITL80409 VITL80410 VITL80411 VITL80412 VITL80413
Bottles, wide neck, with screw cap
Flessen Kunststof flessen
LDPE, translucent, with sealing plug and screw cap made of LDPE.
  • Good chemical resistance
  • Good temperature resistance
Wider mouth for simple filling, also ideal for powders and paste-like materials. With eyes on the bottle and the screw cap for sealing.