HPLC columns, Acclaim™ Organic Acid
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
069700 070086 070087 071987 062902 062903
554-4401EA 357 EUR
554-4401 554-4405 554-4406 554-4407 554-4395 554-4396
HPLC columns, Acclaim™ Organic Acid
Kolommen voor chromatografie
These columns offer unparalleled separations of hydrophilic aliphatic and aromatic organic acids. They are compatible with 100% aqueous mobile phases and low pH conditions. They feature a patented polar-embedded stationary phase that is optimised and use-tested for hydroxyl aliphatic organic acid separations.
  • Hydrolytic stability at low pH conditions, optimum for reversed-phase retention of organic acids
  • Ideal selectivity for separating a wide spectrum of organic acids
  • Excellent peak shapes for organic acids
Particle type: ultra pure, spherical silica
Carbon load: 17%
Surface area: 300 m²/g
pH range: 2 to 8