TLC plates, polyamide-6, polyester sheets, POLYGRAM®
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
803023 803022 803012 803013
552-0153EA 244 EUR
552-0153 552-1292 552-0151 552-0152
TLC plates, polyamide-6, polyester sheets, POLYGRAM®
TLC-platen
Polyamide 6 layers are available as POLYGRAM® polyester sheets with or without fluorescent indicator. The separation mechanism based on hydrogen bonds to amide groups of the polymer matrix as well as on ionic, dipol and electron donor/acceptor interactions.
  • Alternative selectivity to silica 60
Application includes natural compounds, phenols, carboxylic acids, aromatic nitro compounds and especially amino acids.
Polyamide 6: ε-polycaprolactam.