HPTLC plates, nano silica gel 60, modified layers, Nano-SIL C18
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
811052 811054 811062 811064
552-0235EA 414 EUR
552-0235 552-0236 552-0242 552-0243
HPTLC plates, nano silica gel 60, modified layers, Nano-SIL C18
TLC-platen
Octadecyl-modified HPTLC silica glass plates are recommended for alkaloids, amino acids, preservatives, optical brighteners, barbiturates, polycyclic aromatic hydrocarbons (PAH), drugs, peptides, flavonoids, phenols, indole derivatives and steroids.
Base material: Nano-silica 60, specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 toi 10 μm; pH stability 2 to 10.
Modification: Nano-SIL C18-50 has a degree of 50% silanisation; Nano-SIL C18-100 a degree of 100%.
Indicator: Acid-resistant product with a pale blue fluorescence for short-wave UV (254 nm).More Product Information