Copper(II) hexafluoroacetylacetonate hydrate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Hexafluoroacetylacetono Copper(II) Salt, Copper(II) Hexafluoroacetylacetonate, Copper(II) hexafluoro-2,4-pentanedionate hydrate, Bis(hexafluoroacetylacetonato)copper(II) hydrate, Copper(II) hexafluoro-2,4-pentanedionate xhydrate
B25284.03 B25284.06 B25284.14
B25284.03EA 33.5 EUR
B25284.03 B25284.06 B25284.14
Copper(II) hexafluoroacetylacetonate hydrate ≥98%
Copper(II) hexafluoroacetylacetonate hydrate
Purity: 98% CAS No.: 155640-85-0 Molecular Formula: C10H2CuF12O4.xH2O Formula Weight: 477.64(anhy) Melting Point: ca 100° Boiling Point: 220° MDL No.: MFCD00151019
Formule: C₁₀H₂CuF₁₂O₄·H₂O
MW: 477.64 g/mol (anhy)
Kookpunt: 220 °C
Smeltpunt: ∼ 100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00151019
CAS nummer: 155640-85-0
EINECS: 238-850-1