Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](acetanilide)palladium(II)] ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C2387-1G
C2387-200MG
TCIAC2387-1GEA
200
EUR
TCIAC2387-1G
TCIAC2387-200MG
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](acetanilide)palladium(II)] ≥98.0% (by titrimetric analysis)
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](acetanilide)palladium(II)]
This product is unavailable in the U.S.
Formule:
C₃₅H₄₄ClN₃OPd |
CAS nummer:
1228009-69-5 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...