Reinecke's salt ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
423330250 423331000 423335000
ACRO423330250EA 71.5 EUR
ACRO423330250 ACRO423331000 ACRO423335000
Reinecke's salt ACS
Reinecke's salt
Formule: C₄H₁₀CrN₇S₄·1H₂O
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00150925
CAS nummer: 13573-17-6
EINECS: 237-003-3
Order Now
Specification Test Results
Appearance Red Crystalline powder
Infrared spectrum Conforms
Gravimetry ≥93.0 %
Sensitivity passes test
Insoluble matter ≤0.05 % (in dilute HCl)