4-Aminostilbene ≥97.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A0854-25G
A0854-5G
A0854-1G
TCIAA0854-25GEA
1600
EUR
TCIAA0854-25G
TCIAA0854-5G
TCIAA0854-1G
4-Aminostilbene ≥97.0% (by HPLC, titration analysis)
4-Aminostilbene
Formule:
C₁₄H₁₃N |
MDL Nummer:
MFCD00025377 CAS nummer: 834-24-2 |
|
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 150.0 to 153.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...