3,5-Dibromo-1-(trimethylsilyl)benzene ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D2724-25G D2724-5G
TCIAD2724-25GEA 846 EUR
TCIAD2724-25G TCIAD2724-5G
3,5-Dibromo-1-(trimethylsilyl)benzene ≥97.0% (by GC)
3,5-Dibromo-1-(trimethylsilyl)benzene
Formule: C₉H₁₂Br₂Si
MDL Nummer: MFCD03844787
CAS nummer: 17878-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd560a904a1d670b759833e69e797e0d5ce10f90&vid=dd560a904a1d670b759833e69e797e0d5ce10f90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR