2-Acetamidothiazole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A1472-25G A1472-5G
TCIAA1472-25GEA 282 EUR
TCIAA1472-25G TCIAA1472-5G
2-Acetamidothiazole ≥98.0% (by GC)
2-Acetamidothiazole
Formule: C₅H₆N₂OS
MDL Nummer: MFCD00016892
CAS nummer: 2719-23-5
EINECS: 220-321-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7da5d459340d483a58983e36a3a1f5739fd5c76e&vid=7da5d459340d483a58983e36a3a1f5739fd5c76e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 206.0 to 209.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR