L-Glutamic acid 5-methyl ester

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4002301.0005 4002301.0025 4002301.0100
BACE4002301.0005EA 0 EUR
BACE4002301.0005 BACE4002301.0025 BACE4002301.0100
L-Glutamic acid 5-methyl ester
L-Glutamic acid 5-methyl ester
Formule: C₆H₁₁NO₄
MDL Nummer: MFCD00002632
CAS nummer: 1499-55-4
EINECS: 216-110-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR