Methanesulphonamide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

296110250 296111000
ACRO296110250EA 55.8 EUR
ACRO296110250 ACRO296111000
Methanesulphonamide 98%
Methanesulphonamide
Formule: CH₅NO₂S
Smeltpunt: 88…92 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007940
CAS nummer: 3144-09-0
EINECS: 221-553-6

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 88 to 92 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR