Methanesulphonamide ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M0915-25G M0915-500G M0915-100G
TCIAM0915-25GEA 33 EUR
TCIAM0915-25G TCIAM0915-500G TCIAM0915-100G
Methanesulphonamide ≥98.0% (by total nitrogen basis)
Methanesulphonamide
Formule: CH₅NO₂S
MDL Nummer: MFCD00007940
CAS nummer: 3144-09-0
EINECS: 221-553-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7332a5bc2e0f4b179d3d2f0e7d38d7dc13a213a4&vid=7332a5bc2e0f4b179d3d2f0e7d38d7dc13a213a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystaline
Purity (with Total Nitrogen) min. 98.0 %
Melting point 88.0 to 94.0 deg-C
Solubility in Water almost transparency
Solubility in THF faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR