3-Amino-4-methoxybenzenesulphonic acid ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A0491-25G A0491-500G
TCIAA0491-25GEA 22 EUR
TCIAA0491-25G TCIAA0491-500G
3-Amino-4-methoxybenzenesulphonic acid ≥95.0% (by titrimetric analysis)
3-Amino-4-methoxybenzenesulphonic acid
Formule: C₇H₉NO₄S
MDL Nummer: MFCD00035759
CAS nummer: 98-42-0
EINECS: 202-667-5
UN: 2585
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=671e57889cf504c8ec04b98d6f99be547addbf1e&vid=671e57889cf504c8ec04b98d6f99be547addbf1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (Neutralization titration) min. 95.0% (calcd.on dried substance)
Drying loss max. 5.0 %
Sulfuric acid max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR