5-Amino-2-methoxybenzenesulphonic acid ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A1177-25G
A1177-5G
TCIAA1177-25GEA
189
EUR
TCIAA1177-25G
TCIAA1177-5G
5-Amino-2-methoxybenzenesulphonic acid ≥98.0% (by HPLC, titration analysis)
5-Amino-2-methoxybenzenesulphonic acid
Formule:
C₇H₉NO₄S |
MDL Nummer:
MFCD00035760 CAS nummer: 6470-17-3 EINECS: 229-291-4 |
|
Specification Test Results
Appearance | White to Gray to Red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...