Cyclopropylboronic acid tech. 85%, Technical Grade
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H26254.06
H26254.03
H26254.06EA
254
EUR
H26254.06
H26254.03
Cyclopropylboronic acid tech. 85%, Technical Grade
Cyclopropylboronic acid
Purity: 85%
CAS No.: 411235-57-9
Molecular Formula: C3H7BO2
Formula Weight: 85.90
Storage Temperature: -30°C to -10°C
Melting Point: 90-95°
MDL No.: MFCD04038750
Formule:
C₃H₇BO₂ Smeltpunt: 90…95 °C Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD04038750 CAS nummer: 411235-57-9 |