2-Chloro-6-methylnicotinic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C1762-25G
C1762-5G
TCIAC1762-25GEA
333
EUR
TCIAC1762-25G
TCIAC1762-5G
2-Chloro-6-methylnicotinic acid ≥98.0% (by GC, titration analysis)
2-Chloro-6-methylnicotinic acid
Formule:
C₇H₆ClNO₂ MW: 171.58 g/mol Kookpunt: 163°C |
MDL Nummer:
MFCD00134165 CAS nummer: 30529-70-5 EINECS: 250-229-7 |
|
Specification Test Results
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 151.0 to 155.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...