Ethyl-3-ethoxypropionate ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
E0319-25ML E0319-500ML
TCIAE0319-25MLEA 17.6 EUR
TCIAE0319-25ML TCIAE0319-500ML
Ethyl-3-ethoxypropionate ≥99.0% (by GC)
Ethyl-3-ethoxypropionate
Formule: C₇H₁₄O₃
Kookpunt: 170°C
Vlampunt: 49°C
MDL Nummer: MFCD00051356
CAS nummer: 763-69-9
EINECS: 212-112-9
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44c5cc8813909e99a7e4a5fb9df97e578902696b&vid=44c5cc8813909e99a7e4a5fb9df97e578902696b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %