Dinonyl phthalate (mixture of isomers) ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Phthalic acid dinonyl ester

P0303-500ML P0303-25ML
TCIAP0303-500MLEA 345 EUR
TCIAP0303-500ML TCIAP0303-25ML
Dinonyl phthalate (mixture of isomers) ≥95.0% (by GC)
Dinonyl phthalate (mixture of isomers)
Formule: C₂₆H₄₂O₄
Kookpunt: 245°C
Vlampunt: 220°C
MDL Nummer: MFCD00036237
CAS nummer: 84-76-4
EINECS: 201-560-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82c9814d4212b4f3570f8abe79381429e18a092e&vid=b5150516ba92748d33d0c8a021bf436484e0f40c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) max. 95.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR