6-Fluoro-o-toluidine ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
F0817-5G
F0817-1G
TCIAF0817-5GEA
174
EUR
TCIAF0817-5G
TCIAF0817-1G
6-Fluoro-o-toluidine ≥98.0% (by titrimetric analysis)
6-Fluoro-o-toluidine
Formule:
C₇H₈FN Kookpunt: 89°C Vlampunt: 77°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD06658252 CAS nummer: 443-89-0 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...