2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Isopropyl pinacolyl borate, 4,4,5,5-tetramethyl-2-(propan-2-yloxy)-1,3,2-dioxaborolane

I0658-25G I0658-5G I0658-100G
TCIAI0658-25GEA 62.7 EUR
TCIAI0658-25G TCIAI0658-5G TCIAI0658-100G
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formule: C₉H₁₉BO₃
Kookpunt: 80°C
Vlampunt: 37°C
MDL Nummer: MFCD00192241
CAS nummer: 61676-62-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac576071000bc716e872ab65e949b7b33c22bc33&vid=ac576071000bc716e872ab65e949b7b33c22bc33&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR