Tetrabutylammonium hydroxide 55% (w/w) in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
16198.18
16198.30
16198.A1
16198.18EA
67.7
EUR
16198.18
16198.30
16198.A1
Tetrabutylammonium hydroxide 55% (w/w) in aqueous solution
Tetrabutylammonium hydroxide
CAS No.: 2052-49-5
Molecular Formula: C16H37NO
Formula Weight: 259.48
Flash Point: None
Physical Form: Liquid
Density: 0.97
Refractive Index: 1.435
Sensitivity: Air Sensitive
Appearance: Colourless to light yellow
pH: 14
Beilstein Reference: 3571228
UN No.: 3267
MDL No.: MFCD00009425
As a reagent for ion-pair extractions
Formule:
C₁₆H₃₇NO MW: 259.48 g/mol Smeltpunt: 29.85 Dichtheid: 0.97 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009425 CAS nummer: 2052-49-5 EINECS: 218-147-6 UN: 3267 ADR: 8 II |