4'-Amino-2'-methylacetanilide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB06485DA BTB06485EA
MAYBBTB06485DAEA 0 EUR
MAYBBTB06485DA MAYBBTB06485EA
4'-Amino-2'-methylacetanilide
4'-Amino-2'-methylacetanilide
Formule: C₉H₁₂N₂O
MDL Nummer: MFCD00276633
CAS nummer: 56891-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR