N,N-Diisopropylethylenediamine ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D0006-25ML D0006-500ML
TCIAD0006-25MLEA 52.8 EUR
TCIAD0006-25ML TCIAD0006-500ML
N,N-Diisopropylethylenediamine ≥97.0% (by GC, titration analysis)
N,N-Diisopropylethylenediamine
Formule: C₈H₂₀N₂
Kookpunt: 170°C
Vlampunt: 54°C
MDL Nummer: MFCD00015015
CAS nummer: 121-05-1
EINECS: 204-447-4
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afe301794f42fd4cb7d2e73825f734e1ed25a6b6&vid=afe301794f42fd4cb7d2e73825f734e1ed25a6b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR