Zirconium standard solution, 10000 mg/L Zr in 5% nitric acid with 2% hydrofluoric acid standard for ICP
Leverancier: CPACHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C170.5N2FP.L25 C170.5N2FP.L5
ACSDC170.5N2FP.L25EA 529 EUR
ACSDC170.5N2FP.L25 ACSDC170.5N2FP.L5
Zirconium standard solution, 10000 mg/L Zr in 5% nitric acid with 2% hydrofluoric acid standard for ICP
Standaarden Kationische standaarden Zirkonium standaarden