Zirconium standard solution, 10000 mg/L Zr in 5% hydrochloric acid with 2% hydrofluoric acid standard for ICP
Leverancier: ACTIVE MOTIF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C1705C2FP.L1 C1705C2FP.L25 C1705C2FP.L5
ACSDC1705C2FP.L1EA 309 EUR
ACSDC1705C2FP.L1 ACSDC1705C2FP.L25 ACSDC1705C2FP.L5
Zirconium standard solution, 10000 mg/L Zr in 5% hydrochloric acid with 2% hydrofluoric acid standard for ICP
Standaarden Kationische standaarden Zirkonium standaarden