Tin standard solution, 10000 mg/L Sn in dil. nitric acid with 2% hydrofluoric acid standard for ICP
Leverancier: ACTIVE MOTIF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C155.2N2FP.L1 C155.2N2FP.L25 C155.2N2FP.L5
ACSDC155.2N2FP.L1EA 192 EUR
ACSDC155.2N2FP.L1 ACSDC155.2N2FP.L25 ACSDC155.2N2FP.L5
Tin standard solution, 10000 mg/L Sn in dil. nitric acid with 2% hydrofluoric acid standard for ICP
Standaarden Kationische standaarden Tin standaarden