2-Fluoro-3-nitrotoluene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
F0661-25G F0661-5G
TCIAF0661-25GEA 149 EUR
TCIAF0661-25G TCIAF0661-5G
2-Fluoro-3-nitrotoluene ≥98.0% (by GC)
2-Fluoro-3-nitrotoluene
Formule: C₇H₆FNO₂
Kookpunt: 111°C
Smeltpunt: 19°C
MDL Nummer: MFCD03412242
CAS nummer: 437-86-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b2ded5c35b13ab26626c3704fc8006e8111b67b&vid=1b2ded5c35b13ab26626c3704fc8006e8111b67b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 98.0 %