Methyl-5-chlorosalicylate
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Methyl 5-chloro-2-hydroxybenzoate
RF01268.1 RF01268EA
MAYBRF01268.1EA 0 EUR
MAYBRF01268.1 MAYBRF01268EA
Methyl-5-chlorosalicylate
Methyl-5-chlorosalicylate
Formule: C₈H₇ClO₃
MW: 186.60 g/mol
MDL Nummer: MFCD00016461
CAS nummer: 4068-78-4
EINECS: 223-778-5