Palladium standard solution, 10,000 mg/L Pd in 10% nitric acid standard for ICP
Leverancier: ACTIVE MOTIF
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C142.10NP.L1
C142.10NP.L25
C142.10NP.L5
ACSDC142.10NP.L1EA
1060
EUR
ACSDC142.10NP.L1
ACSDC142.10NP.L25
ACSDC142.10NP.L5
Palladium standard solution, 10,000 mg/L Pd in 10% nitric acid standard for ICP
Standaarden
Kationische standaarden
Palladium standaarden