1-Butyl-1-phenylhydrazine ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B2243-25G B2243-5G
TCIAB2243-25GEA 437 EUR
TCIAB2243-25G TCIAB2243-5G
1-Butyl-1-phenylhydrazine ≥98.0% (by titrimetric analysis)
1-Butyl-1-phenylhydrazine
Formule: C₁₀H₁₆N₂
Kookpunt: 105°C
MDL Nummer: MFCD03844777
CAS nummer: 61715-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e03946a6ce7d54ec8f8576a6a53468e0feab331&vid=9e03946a6ce7d54ec8f8576a6a53468e0feab331&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR