3-(Pentafluorothio)aniline ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
3-Aminophenylsulphurpentafluoride, 3-Aminophenylsulphur pentafluoride
H35190.03
H35190.05
H35190.06
H35190.03EA
66.7
EUR
H35190.03
H35190.05
H35190.06
3-(Pentafluorothio)aniline ≥97%
3-(Pentafluorothio)aniline
Formule:
C₆H₆F₅NS Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00674199 CAS nummer: 2993-22-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...