Cyclopropanesulphonyl chloride ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C2187-1G
C2187-5G
TCIAC2187-1GEA
37.4
EUR
TCIAC2187-1G
TCIAC2187-5G
Cyclopropanesulphonyl chloride ≥97.0% (by GC, titration analysis)
Cyclopropanesulphonyl chloride
Formule:
C₃H₅ClO₂S Kookpunt: 60°C Vlampunt: 96°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD01631933 CAS nummer: 139631-62-2 UN: 3265 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...