Bismuth tin alloy (58:42 w%), eutectic ≥99.95% (metals basis), lumps
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12480.14
12480.22
12480.36
12480.14EA
50.9
EUR
12480.14
12480.22
12480.36
Bismuth tin alloy (58:42 w%), eutectic ≥99.95% (metals basis), lumps
Bismuth tin alloy (58:42 w%), eutectic
Purity: 99.95%
Molecular Formula: Bi:Sn; 58:42 wt%
Melting Point: 138°
MDL No.: MFCD00143771