Chlooraceetamide ≥98.0% (by GC) for biochemical research
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-Chloroacetamide
C2536-25G
C2536-5G
TCIAC2536-25GEA
36.3
EUR
TCIAC2536-25G
TCIAC2536-5G
Chlooraceetamide ≥98.0% (by GC) for biochemical research
Chlooraceetamide
Formule:
C₂H₄ClNO Kookpunt: 225°C Smeltpunt: 115…117 °C Dichtheid: 1,6 g/cm³ (20 °C) Vlampunt: 170 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008027 CAS nummer: 79-07-2 EINECS: 201-174-2 UN: 2811 ADR: 6.1,III Merck Index: 14,02110 |
|
Specification Test Results
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Protein impurity test (SDS-PAGE) | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...