3-Hydroxypropiononitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C0450-25ML C0450-100ML
TCIAC0450-25MLEA 27.5 EUR
TCIAC0450-25ML TCIAC0450-100ML
3-Hydroxypropiononitrile ≥98.0% (by GC)
3-Hydroxypropiononitrile
Formule: C₃H₅NO
Kookpunt: 221°C
Smeltpunt: –46 °C
Dichtheid: 1,05 g/cm³ (20 °C)
Vlampunt: 137°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002826
CAS nummer: 109-78-4
EINECS: 203-704-8
Merck Index: 14,03794
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38a2b321fec732a7ef21a5a66a116b4ebf4466c6&vid=38a2b321fec732a7ef21a5a66a116b4ebf4466c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR