2-Butynoic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T1199-1G T1199-25G T1199-5G
TCIAT1199-1GEA 28.6 EUR
TCIAT1199-1G TCIAT1199-25G TCIAT1199-5G
2-Butynoic acid ≥98.0% (by GC, titration analysis)
2-Butynoic acid
Formule: C₄H₄O₂
Kookpunt: 203°C
Smeltpunt: 78 °C
Dichtheid: 0,9641 g/cm³ (20 °C)
MDL Nummer: MFCD00004363
CAS nummer: 590-93-2
EINECS: 209-695-7
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb758fecbed849a92121ce27d9f95cb04253d789&vid=bb758fecbed849a92121ce27d9f95cb04253d789&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 75.0 to 78.0 deg-C