(4-Aminophenyl)acetic acid ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI EUROPE
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
TCIAA0393-25GEA
97.9
EUR
TCIAA0393-25G
(4-Aminophenyl)acetic acid ≥98.0% (by HPLC, titration analysis)
(4-Aminophenyl)acetic acid
Formule:
(C₆H₄(NH₂))CH₂COOH MW: 151,16 g/mol Smeltpunt: 199…202 °C |
MDL Nummer:
MFCD00007916 CAS nummer: 1197-55-3 EINECS: 214-828-7 Merck Index: 13,00462 |
Specification Test Results
Appearance | White to Amber powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...