18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1,4,7,10,13,16-Hexaoxacyclooctadecane, 18-Crown-6
C0860-100G
C0860-25G
C0860-5G
TCIAC0860-100GEA
151
EUR
TCIAC0860-100G
TCIAC0860-25G
TCIAC0860-5G
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥98.0% (by GC)
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)
Formule:
C₁₂H₂₄O₆ Kookpunt: 190°C Smeltpunt: 37…39 °C Dichtheid: 1,237 g/cm³ (25 °C) Vlampunt: 109 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00005113 CAS nummer: 17455-13-9 EINECS: 241-473-5 Merck Index: 14,02602 |
|
Specification Test Results
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Melting point | 39.0 to 43.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...