2-Bromo-5-fluoroanisole ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Bromo-4-fluoro-2-methoxybenzene
B3875-5G B3875-25G
TCIAB3875-5GEA 88 EUR
TCIAB3875-5G TCIAB3875-25G
2-Bromo-5-fluoroanisole ≥95.0% (by GC)
2-Bromo-5-fluoroanisole
Formule: C₇H₆BrFO
Kookpunt: 208°C
Vlampunt: 90°C
MDL Nummer: MFCD04973752
CAS nummer: 450-88-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71fd22de4837c9d99ba95e642264b1a600d615cf&vid=71fd22de4837c9d99ba95e642264b1a600d615cf&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %