2,5-Dimethyl-2,4-hexadiene ≥96.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Stabilisatie: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
D0735-25ML D0735-500ML
TCIAD0735-25MLEA 23.1 EUR
TCIAD0735-25ML TCIAD0735-500ML
2,5-Dimethyl-2,4-hexadiene ≥96.0% (by GC) stabilised
2,5-Dimethyl-2,4-hexadiene
Formule: C₈H₁₄
Kookpunt: 135°C
Smeltpunt: 11 °C
Dichtheid: 0,7625 g/cm³ (20 °C)
Vlampunt: 25°C
MDL Nummer: MFCD00008904
CAS nummer: 764-13-6
EINECS: 212-115-5
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1895bdc88be843d5a70845c9eaf35f4b28c6782&vid=b1895bdc88be843d5a70845c9eaf35f4b28c6782&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %