Dipentyl sulphide ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Dipentylsulphane
A0673-25G A0673-5G
TCIAA0673-25GEA 305 EUR
TCIAA0673-25G TCIAA0673-5G
Dipentyl sulphide ≥97.0% (by GC)
Dipentyl sulphide
Formule: C₁₀H₂₂S
MW: 174.35 g/mol
Smeltpunt: -51.3
Vlampunt: 85°C
MDL Nummer: MFCD00027275
CAS nummer: 872-10-6
EINECS: 212-820-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dfff3070d707561494b72e196d78a1b08e3b845&vid=9dfff3070d707561494b72e196d78a1b08e3b845&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %